accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24598-62-7

IUPAC Name: osmium(4+) diammonium hexabromide
Synonyms: osmium(4+) diammonium hexabromide
Molecular Weight (g/mol): 705.73
Molecular Formula: Br6H8N2Os
InChi Key: ZPHFWWAVHILXPL-UHFFFAOYSA-J
SMILES: [NH4+].[NH4+].[Br-].[Br-].[Br-].[Br-].[Br-].[Br-].[Os+4]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results