accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OO

CAS RN 2461-15-6

IUPAC Name: 2-{[(2-ethylhexyl)oxy]methyl}oxirane
Synonyms: 2-{[(2-ethylhexyl)oxy]methyl}oxirane
Molecular Weight (g/mol): 186.3
Molecular Formula: C11H22O2
InChi Key: BBBUAWSVILPJLL-UHFFFAOYNA-N
SMILES: CCCCC(CC)COCC1CO1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)