accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2461-80-5

IUPAC Name: 2-[(2-oxo-2-phenylethyl)sulfanyl]-1-phenylethan-1-one
Synonyms: 2-[(2-oxo-2-phenylethyl)sulfanyl]-1-phenylethanone
Molecular Weight (g/mol): 270.35
Molecular Formula: C16H14O2S
InChi Key: LHSKVFSCEMNSEC-UHFFFAOYSA-N
SMILES: O=C(CSCC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results