accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24621-21-4

IUPAC Name: niobioamine
Synonyms: niobium nitride
Molecular Weight (g/mol): 106.91
Molecular Formula: NNb
InChi Key: CFJRGWXELQQLSA-UHFFFAOYSA-N
SMILES: N#[Nb]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results