accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2466-76-4

IUPAC Name: 1-(1H-imidazol-1-yl)ethan-1-one
Synonyms: acetylimidazole
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: VIHYIVKEECZGOU-UHFFFAOYSA-N
SMILES: CC(=O)N1C=CN=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)