accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 24672-76-2

IUPAC Name: 9,10-bis(4-methoxyphenyl)anthracene
Synonyms: 9,10-bis(4-methoxyphenyl)anthracene
Molecular Weight (g/mol): 390.48
Molecular Formula: C28H22O2
InChi Key: KTYCXBAOXVVIMM-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=C(OC)C=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results