accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHO

CAS RN 24677-78-9

IUPAC Name: 2,3-dihydroxybenzaldehyde
Synonyms: 2,3-dihydroxybenzaldehyde o-pyrocatechualdehyde
Molecular Weight (g/mol): 138.12
Molecular Formula: C7H6O3
InChi Key: IXWOUPGDGMCKGT-UHFFFAOYSA-N
SMILES: OC1=CC=CC(C=O)=C1O
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (1)
  • (4)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (4)
  • (5)
  • (2)
  • (7)
  • (1)
  • (4)
  • (4)
  • (4)
  • (2)