accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24680-50-0

H3COO(E)

CAS RN 24680-50-0

IUPAC Name: (2E)-3-(4-methoxyphenyl)prop-2-enal
Synonyms: 4-methoxycinnamaldehyde P-methoxycinnamaldehyde
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: AXCXHFKZHDEKTP-NSCUHMNNSA-N
SMILES: COC1=CC=C(\C=C\C=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results