accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOOHO

CAS RN 247027-79-8

IUPAC Name: 3,5-bis(benzyloxy)-2-[(benzyloxy)methyl]-6-(4-methoxyphenoxy)oxan-4-ol
Synonyms: 3,5-bis(benzyloxy)-2-[(benzyloxy)methyl]-6-(4-methoxyphenoxy)oxan-4-ol
Molecular Weight (g/mol): 556.66
Molecular Formula: C34H36O7
InChi Key: MKHXZYRDXXUJMR-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(COCC3=CC=CC=C3)C(OCC3=CC=CC=C3)C(O)C2OCC2=CC=CC=C2)C=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)