accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOH2N

CAS RN 247089-85-6

IUPAC Name: 4-methylbenzene-1-sulfinamide
Synonyms: 4-methylbenzenesulfinamide
Molecular Weight (g/mol): 155.22
Molecular Formula: C7H9NOS
InChi Key: YNJDSRPIGAUCEE-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)