accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 247113-86-6

OHO(S)FFFNHOO

CAS RN 247113-86-6

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[4-(trifluoromethyl)phenyl]propanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-[4-(trifluoromethyl)phenyl]propanoic acid
Molecular Weight (g/mol): 455.43
Molecular Formula: C25H20F3NO4
InChi Key: YMEGJWTUWMVZPD-QFIPXVFZSA-N
SMILES: OC(=O)[C@H](CC1=CC=C(C=C1)C(F)(F)F)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)