accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24715-90-0

IUPAC Name: 4-(pyrrolidin-1-yl)butan-1-amine
Synonyms: 4-(pyrrolidin-1-yl)butan-1-amine
Molecular Weight (g/mol): 142.25
Molecular Formula: C8H18N2
InChi Key: LSDYCEIPEBJKPT-UHFFFAOYSA-N
SMILES: NCCCCN1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results