accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3H3CH3CH3CNCH3H3CH3COSOOO

CAS RN 2472-88-0

IUPAC Name: bis(tetrabutylazanium) sulfate
Synonyms: bis(tetrabutylammonium ion) sulfate
Molecular Weight (g/mol): 581
Molecular Formula: C32H72N2O4S
InChi Key: ZXUCBXRTRRIBSO-UHFFFAOYSA-L
SMILES: [O-]S([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results