accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 247234-41-9

IUPAC Name: N''-[4-(morpholin-4-yl)phenyl]guanidine
Synonyms: N''-[4-(morpholin-4-yl)phenyl]guanidine
Molecular Weight (g/mol): 220.28
Molecular Formula: C11H16N4O
InChi Key: ZVZJREQBRCRGLM-UHFFFAOYSA-N
SMILES: NC(N)=NC1=CC=C(C=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results