accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2473-01-0

IUPAC Name: 1-chlorononane
Synonyms: 1-chlorononane
Molecular Weight (g/mol): 162.7
Molecular Formula: C9H19Cl
InChi Key: RKAMCQVGHFRILV-UHFFFAOYSA-N
SMILES: CCCCCCCCCCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results