accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOOH3CH3CCH3H3CCH3

CAS RN 24748-23-0

IUPAC Name: 3,6,9-triethyl-3,6,9-trimethyl-1,2,4,5,7,8-hexaoxonane
Synonyms: 3,6,9-triethyl-3,6,9-trimethyl-1,2,4,5,7,8-hexaoxonane
Molecular Weight (g/mol): 264.32
Molecular Formula: C12H24O6
InChi Key: KVWLLOIEGKLBPA-UHFFFAOYSA-N
SMILES: CCC1(C)OOC(C)(CC)OOC(C)(CC)OO1
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)