accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CS

CAS RN 24769-39-9

IUPAC Name: 2-decylthiophene
Synonyms: 2-decylthiophene
Molecular Weight (g/mol): 224.41
Molecular Formula: C14H24S
InChi Key: GOTHKCARNFTUSW-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC1=CC=CS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results