accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24772-51-8

IUPAC Name: aluminium(3+) 1-ethoxy-1,3-dioxobutan-2-ide bis(butan-2-olate)
Synonyms: aluminium(3+) 1-ethoxy-1,3-dioxobutan-2-ide bis(butan-2-olate)
Molecular Weight (g/mol): 302.35
Molecular Formula: C14H27AlO5
InChi Key: ATBLFWWYGKIKRM-UHFFFAOYNA-N
SMILES: [Al+3].CCC(C)[O-].CCC(C)[O-].CCOC(=O)[CH-]C(C)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)