accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2478-38-8

IUPAC Name: 1-(4-hydroxy-3,5-dimethoxyphenyl)ethan-1-one
Synonyms: acetosyringone acetosyringenin
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: OJOBTAOGJIWAGB-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1O)C(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (7)
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (3)
  • (3)
  • (4)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (3)
  • (7)
  • (2)
  • (8)
  • (3)
  • (3)
  • (4)
  • (5)