accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3NNNNH3CH3CCH3BrBr

CAS RN 247902-48-3

IUPAC Name: 1-tert-butyl-3-[(1-tert-butyl-1H-imidazol-3-ium-3-yl)methyl]-1H-imidazol-3-ium dibromide
Synonyms: 3-tert-butyl-1-[(3-tert-butylimidazol-1-ium-1-yl)methyl]imidazol-1-ium dibromide
Molecular Weight (g/mol): 422.21
Molecular Formula: C15H26Br2N4
InChi Key: WKZMWHMEXVUXBS-UHFFFAOYSA-L
SMILES: [Br-].[Br-].CC(C)(C)N1C=C[N+](C[N+]2=CN(C=C2)C(C)(C)C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results