accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOCH3OH

CAS RN 2480-86-6

IUPAC Name: 4-(1-hydroxyethyl)-2-methoxyphenol
Synonyms: 4-(1-hydroxyethyl)-2-methoxyphenol
Molecular Weight (g/mol): 168.19
Molecular Formula: C9H12O3
InChi Key: BDRRAMWDUCXAKG-UHFFFAOYNA-N
SMILES: COC1=CC(=CC=C1O)C(C)O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)