accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONHHNOOHOO

CAS RN 2480-93-5

IUPAC Name: 5-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Synonyms: 5-{[(benzyloxy)carbonyl]amino}-2-[(tert-butoxycarbonyl)amino]pentanoic acid
Molecular Weight (g/mol): 366.41
Molecular Formula: C18H26N2O6
InChi Key: QYYCZJUFHDLLOJ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)