accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24806-57-3

IUPAC Name: dimethyl(2-oxo-2-phenylethyl)sulfanium; tetrafluoroboranuide
Synonyms: dimethyl(2-oxo-2-phenylethyl)sulfanium tetrafluoroborate
Molecular Weight (g/mol): 268.08
Molecular Formula: C10H13BF4OS
InChi Key: NYDZUKNJLPHOOF-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.C[S+](C)CC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results