CAS RN 2481-09-6
CAS RN 2481-09-6
IUPAC Name:
hydrogen (2R)-2-amino-3-(tert-butylsulfanyl)propanoic acid chloride
Synonyms:
hydrogen (2R)-2-amino-3-(tert-butylsulfanyl)propanoic acid chloride
Molecular Weight (g/mol):
213.72
Molecular Formula:
C7H16ClNO2S
InChi Key:
MHBMYFJKEBCMDR-JEDNCBNOSA-N
SMILES:
[H+].[Cl-].CC(C)(C)SC[C@H](N)C(O)=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals S-tert-Butyl-L-cysteine hydrochloride, 98%
CAS: 2481-09-6 MDL Number: MFCD00037426