accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2483-46-7

IUPAC Name: (2S)-2,6-bis({[(tert-butoxy)carbonyl]amino})hexanoic acid
Synonyms: (2S)-2,6-bis[(tert-butoxycarbonyl)amino]hexanoic acid
Molecular Weight (g/mol): 346.42
Molecular Formula: C16H30N2O6
InChi Key: FBVSXKMMQOZUNU-LDGXTIHJNA-N
SMILES: CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)