CAS RN 24849-48-7
IUPAC Name:
(2E)-4-(4-hydroxyphenyl)-4-oxobut-2-enoate
Synonyms:
(2E)-4-(4-hydroxyphenyl)-4-oxobut-2-enoate
Molecular Weight (g/mol):
191.16
Molecular Formula:
C10H7O4
InChi Key:
LQCBGQDCRBYBMK-AATRIKPKSA-M
SMILES:
OC1=CC=C(C=C1)C(=O)\C=C\C([O-])=O
1
–
1
of
1
results
1
Promotions Available
4-Hydroxybenzoylacrylic acid, 97%
4-Hydroxybenzoylacrylic acid, 97%
CAS: 24849-48-7 Molecular Formula: C10H7O4 Molecular Weight (g/mol): 191.16 MDL Number: MFCD00460607 InChI Key: LQCBGQDCRBYBMK-AATRIKPKSA-M IUPAC Name: (2E)-4-(4-hydroxyphenyl)-4-oxobut-2-enoate SMILES: OC1=CC=C(C=C1)C(=O)\C=C\C([O-])=O