accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24850-33-7

IUPAC Name: tributyl(prop-2-en-1-yl)stannane
Synonyms: tributyl(prop-2-en-1-yl)stannane
Molecular Weight (g/mol): 331.13
Molecular Formula: C15H32Sn
InChi Key: YLGRTLMDMVAFNI-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)CC=C
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (2)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (6)
  • (1)
  • (4)
  • (4)
  • (2)
  • (4)
  • (4)