accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClOHO

CAS RN 2486-71-7

IUPAC Name: 4-amino-3-chlorobenzoic acid
Synonyms: 4-amino-3-chlorobenzoic acid
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: YIYBPEDZAUFQLO-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1Cl)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results