CAS RN 24864-19-5
IUPAC Name:
4-{[1,1'-biphenyl]-4-yl}-2-methyl-1,3-thiazole
Synonyms:
4-{[1,1'-biphenyl]-4-yl}-2-methyl-1,3-thiazole
Molecular Weight (g/mol):
251.35
Molecular Formula:
C16H13NS
InChi Key:
OKOZRTTWERNRQS-UHFFFAOYSA-N
SMILES:
CC1=NC(=CS1)C1=CC=C(C=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Sigma Aldrich 2-Methyl-4-(4-biphenyl)thiazole
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.