accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiCH3SiCH3H3C

CAS RN 2488-01-9

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 192.41
Molecular Formula: C10H16Si2
InChi Key: UHXCHUWSQRLZJS-UHFFFAOYSA-N
SMILES: C[Si](C)C1=CC=C(C=C1)[Si](C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results