accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2488-14-4

IUPAC Name: methyl 2-{[(tert-butoxy)carbonyl]amino}-3-(1H-imidazol-5-yl)propanoate
Synonyms: methyl 2-[(tert-butoxycarbonyl)amino]-3-(3H-imidazol-4-yl)propanoate
Molecular Weight (g/mol): 269.3
Molecular Formula: C12H19N3O4
InChi Key: RWEVEEORUORBAX-UHFFFAOYNA-N
SMILES: COC(=O)C(CC1=CN=CN1)NC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)