accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2488-15-5

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-4-(methylsulfanyl)butanoic acid|BOC-MET-OH
Molecular Weight (g/mol): 249.33
Molecular Formula: C10H19NO4S
InChi Key: IMUSLIHRIYOHEV-ZETCQYMHSA-N
SMILES: CSCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)