accessibility menu, dialog, popup

UserName

Viewing profile as user:
AgH3COO

CAS RN 2489-05-6

IUPAC Name: silver(1+) docosanoate
Synonyms: silver behenate
Molecular Weight (g/mol): 447.45
Molecular Formula: C22H43AgO2
InChi Key: AQRYNYUOKMNDDV-UHFFFAOYSA-M
SMILES: [Ag+].CCCCCCCCCCCCCCCCCCCCCC([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)