accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2489-86-3

CH2

CAS RN 2489-86-3

IUPAC Name: 1-(prop-2-en-1-yl)naphthalene
Synonyms: 1-allylnaphthalene
Molecular Weight (g/mol): 168.24
Molecular Formula: C13H12
InChi Key: RJFCFNWLPJRCLR-UHFFFAOYSA-N
SMILES: C=CCC1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results