accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24909-68-0

IUPAC Name: (9Z,12Z)-octadeca-9,12-dienoyl (9Z,12Z)-octadeca-9,12-dienoate
Synonyms: (9Z,12Z)-octadeca-9,12-dienoyl (9Z,12Z)-octadeca-9,12-dienoate
Molecular Weight (g/mol): 542.89
Molecular Formula: C36H62O3
InChi Key: BDTYMGCNULYACO-MAZCIEHSSA-N
SMILES: CCCCC\C=C/C\C=C/CCCCCCCC(=O)OC(=O)CCCCCCC\C=C/C\C=C/CCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results