accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3(Z)(Z)

CAS RN 24909-72-6

IUPAC Name: (9Z)-octadec-9-enoyl (9Z)-octadec-9-enoate
Synonyms: (9Z)-octadec-9-enoyl (9Z)-octadec-9-enoate
Molecular Weight (g/mol): 546.92
Molecular Formula: C36H66O3
InChi Key: OCNZHGHKKQOQCZ-CLFAGFIQSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OC(=O)CCCCCCC\C=C/CCCCCCCC
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)