accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2491-31-8

IUPAC Name: 1-(2-hydroxyphenyl)-2-phenylethan-1-one
Synonyms: 1-(2-hydroxyphenyl)-2-phenylethanone
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: VGHVJQXWDXRTRJ-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C(=O)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results