CAS RN 2491-36-3
IUPAC Name:
2-bromo-1-(2-hydroxyphenyl)ethan-1-one
Synonyms:
2-bromo-1-(2-hydroxyphenyl)ethanone
Molecular Weight (g/mol):
215.05
Molecular Formula:
C8H7BrO2
InChi Key:
SGPKEYSZPHMVNI-UHFFFAOYSA-N
SMILES:
OC1=CC=CC=C1C(=O)CBr
1
–
1
of
1
results
1
2-Bromo-2'-hydroxyacetophenone, 97%
CAS: 2491-36-3 Molecular Formula: C8H7BrO2 Molecular Weight (g/mol): 215.046 MDL Number: MFCD01727570 InChI Key: SGPKEYSZPHMVNI-UHFFFAOYSA-N PubChem CID: 200671 IUPAC Name: 2-bromo-1-(2-hydroxyphenyl)ethanone SMILES: C1=CC=C(C(=C1)C(=O)CBr)O