accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2491-38-5

IUPAC Name: 2-bromo-1-(4-hydroxyphenyl)ethan-1-one
Synonyms: 2-bromo-1-(4-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: LJYOFQHKEWTQRH-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(=O)CBr
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results