accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2491-74-9

IUPAC Name: N,N-dimethyl-4-[2-(4-nitrophenyl)diazen-1-yl]aniline
Synonyms: N,N-dimethyl-4-[2-(4-nitrophenyl)diazen-1-yl]aniline
Molecular Weight (g/mol): 270.29
Molecular Formula: C14H14N4O2
InChi Key: LSFRFLVWCKLQTO-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results