accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24936-68-3

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C16H14O3)n
InChi Key: —
SMILES: CC(C)(C1=CC=C(-*)C=C1)C1=CC=C(OC(=O)O-*)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)