accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOH2CCH2H2C

CAS RN 24937-78-8

IUPAC Name: but-3-enoic acid; ethene
Synonyms: ethylene/VA copolymer ethylene-vinyl acetate
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: DQXBYHZEEUGOBF-UHFFFAOYSA-N
SMILES: C=C.OC(=O)CC=C
Quantity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results