accessibility menu, dialog, popup

UserName

Viewing profile as user:
∗HH∗FFn

CAS RN 24937-79-9

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C2H2F2)n
InChi Key: —
SMILES: FC(F)(-*)C-*
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)