accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOCOHOHO

CAS RN 24938-37-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 207.2
Molecular Formula: C8H15O6
InChi Key: CODQTHDBFINLJQ-UHFFFAOYSA-N
SMILES: [O]CCO.[O][C](O)CCCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results