accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24939-16-0

IUPAC Name: (1E,6E)-1,7-bis(4-hydroxyphenyl)hepta-1,6-diene-3,5-dione
Synonyms: bisdemethoxycurcumin bisdemethoxy-curcumin bhcmt bis-demethoxycurcumin
Molecular Weight (g/mol): 308.33
Molecular Formula: C19H16O4
InChi Key: PREBVFJICNPEKM-YDWXAUTNSA-N
SMILES: OC1=CC=C(\C=C\C(=O)CC(=O)\C=C\C2=CC=C(O)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results