CAS RN 24947-68-0
IUPAC Name:
[(1S,2S)-2-hydroxycyclohexyl]methanaminium
Synonyms:
[(1S,2S)-2-hydroxycyclohexyl]methanaminium
Molecular Weight (g/mol):
130.21
Molecular Formula:
C7H16NO
InChi Key:
JGKFBZBVCAWDFD-BQBZGAKWSA-O
SMILES:
[NH3+]C[C@@H]1CCCC[C@@H]1O
1
–
1
of
1
results
1
Promotions Available
cis-2-Aminomethyl-1-cyclohexanol hydrochloride, 99%
cis-2-Aminomethyl-1-cyclohexanol hydrochloride, 99%
CAS: 24947-68-0 Molecular Formula: C7H16NO Molecular Weight (g/mol): 130.21 MDL Number: MFCD00143984 InChI Key: JGKFBZBVCAWDFD-BQBZGAKWSA-O Synonym: 1s,2s-2-hydroxycyclohexyl methanaminium PubChem CID: 2724656 IUPAC Name: (1S,2S)-2-(aminomethyl)cyclohexan-1-ol;hydrochloride SMILES: [NH3+]C[C@@H]1CCCC[C@@H]1O