accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2495-25-2

H3COOCH3CH2

CAS RN 2495-25-2

IUPAC Name: tridecyl 2-methylprop-2-enoate
Synonyms: tridecyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 268.44
Molecular Formula: C17H32O2
InChi Key: KEROTHRUZYBWCY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCOC(=O)C(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results