accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3HNNO

CAS RN 2496-15-3

IUPAC Name: 4-{2-[4-(dimethylamino)phenyl]hydrazin-1-ylidene}cyclohexa-2,5-dien-1-one
Synonyms: 4-{2-[4-(dimethylamino)phenyl]hydrazin-1-ylidene}cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 241.29
Molecular Formula: C14H15N3O
InChi Key: XBCKVYYJXKZISB-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(NN=C2C=CC(=O)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results