accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrNBrBr

CAS RN 24964-91-8

IUPAC Name: 4-bromo-N,N-bis(4-bromophenyl)aniline
Synonyms: 4-bromo-N,N-bis(4-bromophenyl)aniline
Molecular Weight (g/mol): 482.01
Molecular Formula: C18H12Br3N
InChi Key: ZRXVCYGHAUGABY-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N(C1=CC=C(Br)C=C1)C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results