accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3C(Z)

CAS RN 2497-21-4

IUPAC Name: (4Z)-hex-4-en-3-one
Synonyms: (4Z)-hex-4-en-3-one
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: FEWIGMWODIRUJM-HYXAFXHYSA-N
SMILES: CCC(=O)\C=C/C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results